Merck
Heterozygote Wdr36-deficient mice do not develop glaucoma.
Experimental eye research (2014-09-28)
Martin Gallenberger, Markus Kroeber, Loreen März, Marcus Koch, Rudolf Fuchshofer, Barbara M Braunger, Takeshi Iwata, Ernst R Tamm
ABSTRACT
There is an ongoing controversy regarding the role of WDR36 sequence variants in the pathogenesis of primary open-angle glaucoma (POAG). WDR36 is a nucleolar protein involved in the maturation of 18S rRNA. The function of WDR36 is essential as homozygous Wdr36-deficient mouse embryos die before reaching the blastocyst stage. Here we provide a detailed analysis of the phenotype of heterozygous Wdr36-deficient mice. Loss of one Wdr36 allele causes a substantial reduction in the expression of Wdr36 mRNA. In the eyes of Wdr36(+/-) animals, the structure of the tissues involved in aqueous humor circulation and of the optic nerve head are not different from that of control littermates. In addition, one-year-old Wdr36(+/-) animals do not differ from wild-type animals with regards to intraocular pressure and number of optic nerve axons. The susceptibility of retinal ganglion cells to excitotoxic damage induced by NMDA is similar in Wdr36(+/-) and wild-type animals. Moreover, the amount of optic nerve axonal damage induced by high IOP is not different between Wdr36(+/-) and wild-type mice. Transgenic overexpression of mutated Del605-607 Wdr36 in Wdr36(+/-) animals does not cause changes in the number of optic nerve axons or susceptibility to excitotoxic damage. In addition, analysis of 18S rRNA maturation in Del605-607 Wdr36(+/-) or Wdr36(+/-) mice does not show obvious differences in rRNA processing or in the amounts of precursor forms when compared to wild-type animals. Our data obtained in Wdr36(+/-) mice do not support the assumption of a causative role for WDR36 in the pathogenesis of POAG.
Product Description
USP
Maleic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
Maleic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Cresol red, indicator grade, Dye content 95 %
Sigma-Aldrich
N-Methyl-D-aspartic acid, ≥98% (TLC), solid
Sigma-Aldrich
Maleic acid, ReagentPlus®, ≥99% (HPLC)
Supelco
Maleic acid, qNMR standard, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O