Merck
  • MHY-449, a novel dihydrobenzofuro[4,5-b][1,8]naphthyridin-6-one derivative, mediates oxidative stress-induced apoptosis in AGS human gastric cancer cells.
MHY-449, a novel dihydrobenzofuro[4,5-b][1,8]naphthyridin-6-one derivative, mediates oxidative stress-induced apoptosis in AGS human gastric cancer cells.
Oncology reports (2015-05-23)
Seon Hee Kim, Yong Jung Kang, Bokyung Sung, Dong Hwan Kim, Hyun Sook Lim, Hye Rim Kim, Seong Jin Kim, Jeong-Hyun Yoon, Hyung Ryong Moon, Hae Young Chung, Nam Deuk Kim
ABSTRACT
MHY-449 is a novel dihydrobenzofuro[4,5-b][1,8]naphthyridin-6-one derivative designed and synthesized as a potential anticancer agent. The present study aimed to examine the anticancer activity and underlying mechanism of MHY-449. The cell viability assay performed in AGS human gastric carcinoma cells demonstrated that MHY-449 inhibited cell proliferation in a concentration-dependent manner. MHY-449 induced AGS cell death via apoptosis. The underlying molecular mechanism of MHY-449-mediated apoptosis was also investigated. MHY-449 promoted the upregulation of Fas and Fas-ligand, and activation of caspase-8, suggesting the involvement of a Fas-mediated extrinsic pathway in MHY-449-induced apoptosis. In addition, it was found that MHY-449-induced apoptosis was accompanied by the upregulation of Bax, p21(WAF1/CIP1), p27(KIP1), and p53 and suppression of Bcl-2. MHY-449 exposure activated the caspase cascade and subsequent poly(ADP-ribose) polymerase (PARP) cleavage. Furthermore, the pan-caspase inhibitor, Z-VAD-FMK, significantly attenuated MHY-449-induced apoptosis, indicating that the apoptosis was caspase-dependent. Moreover, the apoptogenic effect of MHY-449 was reactive oxygen species (ROS)-dependent. This result was confirmed by the induction of ROS by MHY-449 and by evidence that the scavenging of ROS by N-acetyl-L-cysteine inhibited MHY-449-induced cell death. Taken together, these results demonstrated that MHY-449 triggers apoptosis via caspase activation and ROS production. This result provides a novel mechanistic explanation and a basis for developing this compound as a novel candidate for human cancer therapy.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
4-Nitroaniline, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
DCFHDA, BioReagent, suitable for fluorescence, ≥95% (HPLC)
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)