Merck
Dual Regulatory Role of Polyamines in Adipogenesis.
The Journal of biological chemistry (2015-09-24)
Shirley Brenner, Zippi Bercovich, Yulia Feiler, Rom Keshet, Chaim Kahana
ABSTRACT
Adipogenesis is a complex process, accompanied by a chain of interdependent events. Disruption of key events in this cascade may interfere with the correct formation of adipose tissue. Polyamines were demonstrated necessary for adipogenesis; however, the underlying mechanism by which they act has not been established. Here, we examined the effect of polyamine depletion on the differentiation of 3T3-L1 preadipocytes. Our results demonstrate that polyamines are required early in the adipogenic process. Polyamine depletion inhibited the second division of the mitotic clonal expansion (MCE), and inhibited the expression of PPARγ and C/EBPα, the master regulators of adipogenesis. However, it did not affect the expression of their transcriptional activator, C/EBPβ. Additionally, polyamine depletion resulted in elevation of mRNA and protein levels of the stress-induced C/EBP homologous protein (CHOP), whose dominant negative function is known to inhibit C/EBPβ DNA binding activity. Conditional knockdown of CHOP in polyamine-depleted preadipocytes restored PPARγ and C/EBPα expression, but failed to recover MCE and differentiation. Thus, our results suggest that the need for MCE in the adipogenic process is independent from the requirement for PPARγ and C/EBPα expression. We conclude that de novo synthesis of polyamines during adipogenesis is required for down-regulation of CHOP to allow C/EBPβ activation, and for promoting MCE.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
Acetaldehyde solution, 5 M in THF
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Diphenylamine, ACS reagent, ≥99%
Sigma-Aldrich
Acetaldehyde solution, 50 wt. % in ethanol
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Diphenylamine, ReagentPlus®, 99%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, BioUltra, ≥99%
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material