Merck
  • Differences in histone modifications between slow- and fast-twitch muscle of adult rats and following overload, denervation, or valproic acid administration.
Differences in histone modifications between slow- and fast-twitch muscle of adult rats and following overload, denervation, or valproic acid administration.
Journal of applied physiology (Bethesda, Md. : 1985) (2015-09-26)
Fuminori Kawano, Keisuke Nimura, Saki Ishino, Naoya Nakai, Ken Nakata, Yoshinobu Ohira
ABSTRACT
Numerous studies have reported alterations in skeletal muscle properties and phenotypes in response to various stimuli such as exercise, unloading, and gene mutation. However, a shift in muscle fiber phenotype from fast twitch to slow twitch is not completely induced by stimuli. This limitation is hypothesized to result from the epigenetic differences between muscle types. The main purpose of the present study was to identify the differences in histone modification for the plantaris (fast) and soleus (slow) muscles of adult rats. Genome-wide analysis by chromatin immunoprecipitation followed by DNA sequencing revealed that trimethylation at lysine 4 and acetylation of histone 3, which occurs at transcriptionally active gene loci, was less prevalent in the genes specific to the slow-twitch soleus muscle. Conversely, gene loci specific to the fast-twitch plantaris muscle were associated with the aforementioned histone modifications. We also found that upregulation of slow genes in the plantaris muscle, which are related to enhanced muscular activity, is not associated with activating histone modifications. Furthermore, silencing of muscle activity by denervation caused the displacement of acetylated histone and RNA polymerase II (Pol II) in 5' ends of genes in plantaris, but minor effects were observed in soleus. Increased recruitment of Pol II induced by forced acetylation of histone was also suppressed in valproic acid-treated soleus. Our present data indicate that the slow-twitch soleus muscle has a unique set of histone modifications, which may relate to the preservation of the genetic backbone against physiological stimuli.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
2-Propylpentanoic acid
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri