Merck
  • Growth arrest-specific protein 6 protects against renal ischemia-reperfusion injury.
Growth arrest-specific protein 6 protects against renal ischemia-reperfusion injury.
The Journal of surgical research (2015-07-18)
Matthew D Giangola, Weng-Lang Yang, Salil R Rajayer, Michael Kuncewitch, Ernesto Molmenti, Jeffrey Nicastro, Gene F Coppa, Ping Wang
ABSTRACT
Renal injury caused by ischemia-reperfusion (I/R) often occurs after shock or transplantation. Growth arrest-specific protein 6 (Gas6) is a secreted protein that binds to the TAM-Tyro3, Axl, Mer-family tyrosine kinase receptors, which modulate the inflammatory response and activate cell survival pathways. We hypothesized that Gas6 could have a protective role in attenuating the severity of renal injury after I/R. Adult mice were subjected to 45 min of bilateral renal ischemia. Recombinant mouse Gas6 (rmGas6, 5 μg per mouse) or normal saline (vehicle) was administered intraperitoneally 1 h before ischemia and all subjects were sacrificed at 23 h after I/R for blood and tissue analysis. The expression of protein and messenger RNA (mRNA) was assessed by Western blotting and quantitative polymerase chain reaction, respectively. Treatment with rmGas6 significantly decreased serum levels of creatinine and blood urea nitrogen by 29% and 27%, respectively, improved the renal histologic injury index, and reduced the apoptosis in the kidneys, compared with the vehicle. Renal mRNA levels of interleukin 1β, interleukin 6, tumor necrosis factor α, keratinocyte-derived chemokine and macrophage inflammatory protein 2 were decreased significantly by 99%, 60%, 53%, 58%, and 43%, with rmGas6 treatment, respectively. After I/R, renal I-kappa-B α levels were reduced by 40%, whereas they returned to sham levels with rmGas6 treatment. The mRNA levels of inducible nitric oxide synthase and cyclooxygenase 2 were reduced by 79% and 70%, respectively, whereas the expression of cyclin D1 was increased by 2.1-fold in the rmGas6-treated group, compared with the vehicle. Gas6 suppresses the nuclear factor κB pathway and promotes cell proliferation, leading to the reduction of inflammation and protection of renal injury induced by I/R.
Sigma-Aldrich
Sodium chloride solution, 0.85%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)