Merck
  • Urinary polycyclic aromatic hydrocarbon metabolites as biomarkers of exposure to traffic-emitted pollutants.
Urinary polycyclic aromatic hydrocarbon metabolites as biomarkers of exposure to traffic-emitted pollutants.
Environment international (2015-09-19)
Jicheng Gong, Tong Zhu, Howard Kipen, David Q Rich, Wei Huang, Wan-Ting Lin, Min Hu, Junfeng Jim Zhang
ABSTRACT
1-Nitro-pyrene has been considered a compound specific to diesel combustion emission, while 1- and 2-nitro-napthalene are mainly produced through photochemical conversion of naphthalene released to the atmosphere. Metabolites of these compounds may serve as biomarkers of exposure to traffic related pollutants. We collected urine samples from 111 healthy and non-smoking subjects within (i.e., during the Beijing Olympics) and outside (i.e., before and after the Olympics) a traffic control regime to improve Beijing's air quality. Urines were analyzed for the sum of 1&2-amino-naphthalene (metabolites of 1- and 2-nitro-naphthalene) and 1-amino-pyrene (a metabolite of 1-nitro-pyrene), using an HPLC-fluorescence method. Within the same time periods, PM2.5 mass and constituents were measured, including elemental carbon, sulfate, nitrate, PAHs, carbon monoxide, nitrogen dioxide, sulfur dioxide, ozone, and particle number concentrations. The associations between the urinary metabolites and air pollutants were analyzed using linear mixed-effects models. From the pre- to during-Olympic period, 1&2-amino-naphthalene and 1-hydroxy-pyrene decreased by 23% (p=0.066) and 16% (p=0.049), respectively, while there was no change in 1-amino-pyrene (2% increase, p=0.892). From during- to post-Olympic period, 1&2-amino-naphthalene, 1-amino-pyrene and 1-hydroxy-pyrene concentrations increased by 26% (p=0.441), 37% (p=0.355), and 3% (p=0.868), respectively. Furthermore, 1&2-amino-naphthalene and 1-hydroxy-pyrene were associated with traffic related pollutants in a similar lag pattern. 1-amino-pyrene was associated more strongly with diesel combustion products (e.g. PN and elemental carbon) and not affected by season. Time-lag analyses indicate strongest/largest associations occurred 24-72h following exposure. 1&2-amino-naphthalene and 1-hydroxy-pyrene can be used as a biomarker of exposure to general vehicle-emitted pollutants. More data are needed to confirm 1-amino-pyrene as a biomarker of exposure to diesel combustion emissions. Controlling creatinine as an independent variable in the models will provide a moderate adjusting effect on the biomarker analysis.
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture