Merck
  • Alum and Rainfall Effects on Ionophores in Runoff from Surface-Applied Broiler Litter.
Alum and Rainfall Effects on Ionophores in Runoff from Surface-Applied Broiler Litter.
Journal of environmental quality (2015-10-06)
Sarah A Doydora, Dorcas Franklin, Peizhe Sun, Miguel Cabrera, Aaron Thompson, Kimberly Love-Myers, John Rema, Vaughn Calvert, Spyros G Pavlostathis, Ching-Hua Huang
ABSTRACT
Polyether ionophores, monensin, and salinomycin are commonly used as antiparasitic drugs in broiler production and may be present in broiler litter (bird excreta plus bedding material). Long-term application of broiler litter to pastures may lead to ionophore contamination of surface waters. Because polyether ionophores break down at low pH, we hypothesized that decreasing litter pH with an acidic material such as aluminum sulfate (alum) would reduce ionophore losses to runoff (i.e., monensin and salinomycin concentrations, loads, or amounts lost). We quantified ionophore loss to runoff in response to (i) addition of alum to broiler litter and (ii) length of time between litter application and the first simulated rainfall event. The factorial experiment consisted of unamended (∼pH 9) vs. alum-amended litters (∼pH 6), each combined with simulated rainfall at 0, 2, or 4 wk after litter application. Runoff from alum-amended broiler litter had 33% lower monensin concentration ( < 0.01), 57% lower monensin load ( < 0.01), 48% lower salinomycin concentration ( < 0.01), and 66% lower salinomycin load ( < 0.01) than runoff from unamended broiler litter when averaged across all events of rainfall. Ionophore losses to runoff were also less when rainfall was delayed for 2 or 4 wk after litter application relative to applying rainfall immediately after litter application. While the weather is difficult to predict, our data suggest that ionophore losses in runoff can be reduced if broiler litter applications are made to maximize dry time after application.
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)