Merck
  • Transplantation of engineered organoids enables rapid generation of metastatic mouse models of colorectal cancer.
Transplantation of engineered organoids enables rapid generation of metastatic mouse models of colorectal cancer.
Nature biotechnology (2017-05-02)
Kevin P O'Rourke, Evangelia Loizou, Geulah Livshits, Emma M Schatoff, Timour Baslan, Eusebio Manchado, Janelle Simon, Paul B Romesser, Benjamin Leach, Teng Han, Chantal Pauli, Himisha Beltran, Mark A Rubin, Lukas E Dow, Scott W Lowe
ABSTRACT
Colorectal cancer (CRC) is a leading cause of death in the developed world, yet facile preclinical models that mimic the natural stages of CRC progression are lacking. Through the orthotopic engraftment of colon organoids we describe a broadly usable immunocompetent CRC model that recapitulates the entire adenoma-adenocarcinoma-metastasis axis in vivo. The engraftment procedure takes less than 5 minutes, shows efficient tumor engraftment in two-thirds of mice, and can be achieved using organoids derived from genetically engineered mouse models (GEMMs), wild-type organoids engineered ex vivo, or from patient-derived human CRC organoids. In this model, we describe the genotype and time-dependent progression of CRCs from adenocarcinoma (6 weeks), to local disseminated disease (11-12 weeks), and spontaneous metastasis (>20 weeks). Further, we use the system to show that loss of dysregulated Wnt signaling is critical for the progression of disseminated CRCs. Thus, our approach provides a fast and flexible means to produce tailored CRC mouse models for genetic studies and pre-clinical investigation.
Sigma-Aldrich
Albumin, glycated human, lyophilized powder
Pricing and availability is not currently available.
Sigma-Aldrich
Y-27632, InSolution, ≥95%, reversible, inhibitor of Rho kinases
Pricing and availability is not currently available.