Pozwól nam pomócInchi Code
1S/C14H13NO4S/c1-10-5-7-13(8-6-10)20(18,19)15-12-4-2-3-11(9-12)14(16)17/h2-9,15H,1H3,(H,16,17)
Inchi Key
FEPILVGKCQHGLG-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
97%
Storage temp.
Sealed in dry, room temperature
Country of Origin
CN
Physical Form
Powder or crystals
Still not finding the right product?
Explore all of our products under 3-((4-Methylphenyl)sulfonamido)benzoic acid
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?