Pozwól nam pomócInchi Code
1S/C14H11F3O/c15-14(16,17)13-7-5-12(6-8-13)11-3-1-10(9-18)2-4-11/h1-8,18H,9H2
Inchi Key
YCVFDTRUBSFXSA-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
97%
Storage temp.
Sealed in dry, room temperature
Country of Origin
CN
Physical Form
Solid
Still not finding the right product?
Explore all of our products under (4'-(Trifluoromethyl)-[1,1'-biphenyl]-4-yl)methanol
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?