Pozwól nam pomócInchi Code
1S/C14H16N2/c1-9-5-3-7-11(15)13(9)14-10(2)6-4-8-12(14)16/h3-8H,15-16H2,1-2H3
Inchi Key
MCUUKQCKNKUMBP-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
97%
Storage temp.
Keep in dark place, inert atmosphere, room temperature
Country of Origin
CN
Physical Form
Solid
Still not finding the right product?
Explore all of our products under (R)-6,6'-Dimethyl-[1,1'-biphenyl]-2,2'-diamine
hstatements
Słowo ostrzegawcze
Danger
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?