Skip to Content
MilliporeSigma
Skip To
BDPH9BCD856F
Bide Pharmatech
2-(4-(Dimethylamino)phenyl)-1H-benzo[d]imidazol-5-amine
Synonym(s):
N-[4-(5-amino-1H-benzimidazol-2-yl)phenyl]-N,N-dimethylamine
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Size/SKUAvailabilityPrice
About This Item
Linear Formula:
C15H16N4
CAS Number:
Molecular Weight:
252.32
MDL number:
Product BDPH9BCD856F is currently not available for purchase. Contact Technical Service
Inchi Code
1S/C15H16N4/c1-19(2)12-6-3-10(4-7-12)15-17-13-8-5-11(16)9-14(13)18-15/h3-9H,16H2,1-2H3,(H,17,18)
Inchi Key
MYBHZNWJSKGXNY-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
95%
Storage temp.
Keep in dark place, sealed in dry, room temperature
Country of Origin
CN
Physical Form
Solid
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library