Pozwól nam pomócInchi Code
1S/C9H4ClF3N2/c10-8-14-4-5-3-6(9(11,12)13)1-2-7(5)15-8/h1-4H
Inchi Key
XGKICHJOVNGJBU-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
98%
Storage temp.
Inert atmosphere, 2-8C
Country of Origin
CN
Physical Form
Solid
Still not finding the right product?
Explore all of our products under 2-Chloro-6-(trifluoromethyl)quinazoline
hstatements
signalword
Danger
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?