Pozwól nam pomócInchi Code
1S/C30H48P2/c1-5-15-25(16-6-1)31(26-17-7-2-8-18-26)29-23-13-14-24-30(29)32(27-19-9-3-10-20-27)28-21-11-4-12-22-28/h13-14,23-28H,1-12,15-22H2
Inchi Key
DZNBZPKZOBDXEL-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
98%
Storage temp.
Keep in dark place, inert atmosphere, room temperature
Country of Origin
CN
Physical Form
Solid
Still not finding the right product?
Explore all of our products under 1,2-Bis(dicyclohexylphosphino)benzene
hstatements
signalword
Warning
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?