Pozwól nam pomócInchi Code
1S/C5H9NO3.ClH/c7-5(8)4-3-9-2-1-6-4;/h4,6H,1-3H2,(H,7,8);1H/t4-;/m0./s1
Inchi Key
CWSLARZELUGARZ-WCCKRBBISA-N
Shipping Temperature
Normal
Purity
97%
Storage temp.
Inert atmosphere, room temperature
Country of Origin
CN
Physical Form
White to Pale-yellow to Yellow-brown Solid
Still not finding the right product?
Explore all of our products under (S)-Morpholine-3-carboxylic acid hydrochloride
hstatements
Ostrzeżenia
Słowo ostrzegawcze
Warning
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?