Pozwól nam pomócInchi Code
1S/C8H9N3.2ClH/c1-6(10)8-7(5-9)3-2-4-11-8;;/h2-4,6H,10H2,1H3;2*1H/t6-;;/m1../s1
Inchi Key
DZWVKGOYKLAUDO-QYCVXMPOSA-N
Shipping Temperature
Normal
Purity
97%
Storage temp.
Inert atmosphere, 2-8C
Country of Origin
CN
Physical Form
Solid
Still not finding the right product?
Explore all of our products under (R)-2-(1-Aminoethyl)nicotinonitrile dihydrochloride
hstatements
signalword
Warning
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?