Pozwól nam pomócInchi Code
1S/C4H5N3O2/c5-4-6-1-2(7-4)3(8)9/h1H,(H,8,9)(H3,5,6,7)
Inchi Key
YTYZMZQDGZMTJM-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
97%
Country of Origin
CN
Physical Form
Solid
Storage temp.
Keep in dark place, inert atmosphere, 2-8C
Still not finding the right product?
Explore all of our products under 2-Amino-1H-imidazole-5-carboxylic acid
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?