Pozwól nam pomócInchi Code
1S/C7H4F3NO3/c8-7(9,10)14-6-3-1-5(2-4-6)11(12)13/h1-4H
Inchi Key
UBEIKVUMDBCCRW-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
98%
Storage temp.
Sealed in dry, room temperature
Country of Origin
CN
Physical Form
Solid or Semi-Solid or Liquid or Lump
Still not finding the right product?
Explore all of our products under 1-Nitro-4-(trifluoromethoxy)benzene
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?