Pozwól nam pomócInchi Code
1S/C9H8N2/c10-9-3-1-2-7-6-11-5-4-8(7)9/h1-6H,10H2
Inchi Key
DTVYNUOOZIKEEX-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
98%
Storage temp.
Keep in dark place, sealed in dry, room temperature
Country of Origin
CN
Physical Form
Form: Crystal - Powder / Colour: Pale yellow - Greyish yellow red - Gray
Melting Point
125-128C
Still not finding the right product?
Explore all of our products under Isoquinolin-5-amine
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?