Pozwól nam pomócInchi Code
1S/C8H9NO/c1-6-3-8(5-10)4-7(2)9-6/h3-5H,1-2H3
Inchi Key
OFIMNDTVWZPKIN-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
98%
Country of Origin
CN
Physical Form
Liquid or Solid or Semi-solid
Storage temp.
Inert atmosphere, 2-8C
Still not finding the right product?
Explore all of our products under 2,6-Dimethylisonicotinaldehyde
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?