Pozwól nam pomócInchi Code
1S/C7H3F3INO2/c8-7(9,10)5-2-1-4(12(13)14)3-6(5)11/h1-3H
Inchi Key
GXQIWTSUPAYASC-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
95%
Country of Origin
CN
Physical Form
Pale-yellow to Yellow-brown Solid
Storage temp.
Keep in dark place, sealed in dry, 2-8C
Still not finding the right product?
Explore all of our products under 2-Iodo-4-nitro-1-(trifluoromethyl)benzene
hstatements
Ostrzeżenia
Słowo ostrzegawcze
Warning
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?