Pozwól nam pomócInchi Code
1S/C13H17NO3/c1-13(2,3)14-11(15)9-5-7-10(8-6-9)12(16)17-4/h5-8H,1-4H3,(H,14,15)
Inchi Key
VASLEASMBRLHMM-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
98%
Storage temp.
Sealed in dry, room temperature
Country of Origin
CN
Physical Form
Solid
Still not finding the right product?
Explore all of our products under Methyl 4-(tert-butylcarbamoyl)benzoate
hstatements
Słowo ostrzegawcze
Warning
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?