Przejdź do
A002
NovaSeptum Stojak na worek, pojedyncze próbkowanie, dla 1-portowego uchwytu TC25/NAC18
For use with 1-port holder, TC 25/NAC 18
Synonim(y):
single-sample bag rack, sterile sample bag rack
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
1 ea
Przewidywany termin wysyłki21 października 2026zKuehne + Nagel Sp. z o.o.
1900,00 zł
Informacje o tej pozycji
eCl@ss:
36101790
UNSPSC Code:
41104105
1900,00 zł
Przewidywany termin wysyłki21 października 2026Szczegóły
​
Pomoc techniczna
Pozwól nam pomóc
Quality Segment
material
stainless steel
sterility
non-sterile
product line
NovaSeptum®
packaging
pkg of 1 unit
L × W × H
250 mm (9.8 in.) × 30 mm (1.2 in.) × 260-460 mm (10.2-18.1 in.)
weight
0.5 kg1.1 lb
application(s)
sterile sampling
compatibility
for use with 1-port holder, TC 25/NAC 18
General description
The NovaSeptum® Bag Rack for Single Sampling Units is a stainless steel mechanical support rack designed to hold and organize NovaSeptum® sampling units during aseptic fluid sampling operations. The rack is engineered for compatibility with the 1-port holder in a TC 25/NAC 18 (Tri-Clamp 25 mm / NovAseptic Connector 18 mm) configuration. The rack is fully adjustable, reusable, and available for all standard holders.
Application
  • The NovaSeptum® Bag Rack acts as a structured mechanical support frame that holds and organizes NovaSeptum® sampling units during aseptic fluid sampling operations.
  • It is particularly suited for use in biopharmaceutical manufacturing and cleanroom environments where controlled, contamination-free fluid sampling is required.
Features and Benefits
  • Fully adjustable to accommodate different process setups.
  • The maximum load capacity of the rack goes up to 5 kgs.
  • The structural components are manufactured from stainless steel in compliance with ASTM® 316, while the screws and locking nuts are made from A4-grade stainless steel.
  • A certificate of quality is included in each package for traceability and compliance.
  • Compatible with bag, bottle and conical tube sampling units
Packaging
A complete bag rack unit, including all components, is packaged unassembled in a cardboard box and assigned a part number.
Legal Information
ASTM is a registered trademark of American Society for Testing and Materials
NOVASEPTUM is a registered trademark of Merck KGaA, Darmstadt, Germany
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
A007A001A006
description
For use with 1-port holder, TC 25/NAC 18
description
For use with 1-port holder, TC 25/NAC 18
description
-
description
-
compatibility
for use with 1-port holder, TC 25/NAC 18
compatibility
for use with 1-port holder, TC 25/NAC 18
compatibility
-
compatibility
-
product line
NovaSeptum®
product line
NovaSeptum®
product line
-
product line
-
material
stainless steel
material
stainless steel (Manifold holder: Polyacetal copolymer)
material
-
material
-
L × W × H
250 mm (9.8 in.) × 30 mm (1.2 in.) × 260-460 mm (10.2-18.1 in.)
L × W × H
300 mm (11.8 in.) × 60 mm (2.3 in.) × 240-440 mm (9.4-17.3 in.)
L × W × H
-
L × W × H
-
application(s)
sterile sampling
application(s)
sterile sampling
application(s)
-
application(s)
-
sterility
non-sterile
sterility
non-sterile
sterility
sterile
sterility
sterile
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)