Merck
02252
(−)-Epinephrine
tested according to USP
Synonim(y):
(−)-Adrenalin, (R)-(−)-3,4-Dihydroxy-α-(methylaminomethyl)benzyl alcohol, L-Adrenaline, L-Epinephrine
Informacje o tej pozycji
Wzór liniowy:
(HO)2C6H3CH(OH)CH2NHCH3
Numer CAS:
Masa cząsteczkowa:
183.20
EC Number:
200-098-7
UNSPSC Code:
12352200
PubChem Substance ID:
Beilstein/REAXYS Number:
2368277
MDL number:
Pomoc techniczna
Pozwól nam pomóc
agency
tested according to USP
mp
215 °C (dec.) (lit.)
solubility
H2O: very slightly soluble, NaOH: very soluble, acetone: insoluble, alcohol: very slightly soluble, chloroform: insoluble, diethyl ether: insoluble, oil: insoluble, potassium hydroxide solution: very soluble
application(s)
pharmaceutical (small molecule)
storage temp.
2-8°C
SMILES string
CNC[C@H](O)c1ccc(O)c(O)c1
InChI
1S/C9H13NO3/c1-10-5-9(13)6-2-3-7(11)8(12)4-6/h2-4,9-13H,5H2,1H3/t9-/m0/s1
InChI key
UCTWMZQNUQWSLP-VIFPVBQESA-N
Application
(-)-Epinephrine was used in fertilization medium for rinsing Cumulus-oocyte complexes (COC) during in vitro maturation.
Biochem/physiol Actions
(-)-Epinephrine enhances the IL-6 secretion and bioactivity from isolated perfused rat liver (IPRL).
Adrenoceptor agonist.
Preparation Note
(-)-Epinephrine readily dissolves in aqueous solutions of mineral acids, KOH and NaOH. It is very slightly soluble in water and alcohol but is insoluble in chloroform, ether, acetone, oils as well as in aqueous solutions of ammonia and the alkali carbonates.
Still not finding the right product?
Explore all of our products under (−)-Epinephrine
Piktogramy
Skull and crossbones
Słowo ostrzegawcze
Danger
Kody zagrożeń
Hazard Classifications
Acute Tox. 2 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral
Klasa składowania
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Not applicable
PPE (środki ochrony indywidualnej)
Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)