Zmień widok
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
eCl@ss:
32040501
UNSPSC Code:
41122104
Material:
canted neck , cap (vented), polystyrene , rectangular flask
Sterility:
sterile
Feature:
graduations
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomócmaterial
canted neck , cap (vented), polystyrene , rectangular flask
sterility
sterile
feature
graduations
packaging
case of 100
manufacturer/tradename
Corning 430641
technique(s)
cell culture: suitable
maximum volume
60 mL
surface area
75 cm2
working volume
15-22.5 mL
General description
Available in a variety of sizes, designs and cap styles to meet your needs.
- Manufactured from optically clear virgin polystyrene
- Treated for optimal cell attachment
- Printed with lot numbers for ease in traceability
- 100% integrity tested
- Sterilized by gamma irradiation and certified nonpyrogenic
Application
Legal Information
Corning is a registered trademark of Corning, Inc.
Still not finding the right product?
Explore all of our products under Corning® cell culture flasks
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?