Informacje o tej pozycji
UNSPSC Code:
41122410
NACRES:
NB.43
eCl@ss:
32120613
Manufacturer/tradename:
Benchmark Scientific MS1000-F2
Packaging:
pack of 100 ea
Pomoc techniczna
Pozwól nam pomócpackaging
pack of 100 ea
manufacturer/tradename
Benchmark Scientific MS1000-F2
film W × L
125 mm × 78 mm
General description
Strong Bonding foil seals designed for heat sealing of polypropylene microplates. Ideal for cold storage (-200° to 110°C) and with samples contianing DMSO. Compatible with most standard heat sealers. Pack of 100.
Application
Cold sample storage in microplates
Features and Benefits
Strong bonding for cold storage
Still not finding the right product?
Explore all of our products under Benchmark Foil Seals
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?