Przejdź do
MTCC2230AM
MTC™ Bio ScrewSeal™ Microcentrifuge Tubes
self-standing, amber, capacity 2.0 mL, pkg of 1000 ea (10 x bags 100 ea)
Synonim(y):
conical tube, plastic tube, MTC Bio C2230-AM, sample collection tube, sample receptacle, sample tube
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
Informacje o tej pozycji
NACRES:
NB.15
UNSPSC Code:
41121800
Capacity:
2.0 mL
Material:
amber , high-density polyethylene cap, polypropylene , screw top cap, self-standing
Sterility:
sterile
Packaging:
pkg of 1000 ea (10 x bags 100 ea)
Parameter:
22,000 × g speed
Feature:
graduation
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomóc
material
amber , high-density polyethylene cap, polypropylene , screw top cap, self-standing
sterility
sterile
feature
graduation
packaging
pkg of 1000 ea (10 x bags 100 ea)
manufacturer/tradename
MTC™ Bio C2230-AM
parameter
22,000 × g speed
capacity
2.0 mL
diam. × L
12.8 mm × 47.4 mm
General description
ScrewSeal™ screw-cap microtubes are made from ultra-clear polypropylene, these screw cap microtubes include assembled caps with silicone O-rings that assure a leak-proof seal from -90°C to +121°C. Sterile, RNase, DNase, & pyrogen free and may be centrifuged at up to 22,000×g.
Screw-cap microtubes, 2.0mL, amber color, with O-ring, sterile, molded graduations, amber caps attached, 100 per bag, 1000/case
Features and Benefits
  • Ultra-clear polypropylene
  • Molded or non-graduated options
  • Leak-proof O-ring seal
  • Light-blocking amber available
  • Sterile, RNase, DNase, Pyrogen free
Legal Information
MTC is a trademark of MTC Bio
ScrewSeal is a trademark of MTC Bio
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
MTCC2230MTCC2231WMTCC3150S
description
self-standing, amber, capacity 2.0 mL, pkg of 1000 ea (10 x bags 100 ea)
description
self-standing bottom, clear, capacity 2.0 mL, pack of 1000 ea (10 x bags 100 ea)
description
self-standing, clear, capacity 2.0 mL, pkg of 500 ea (individually wrapped)
description
conical clear bottom, capacity 1.5 mL, pack of 1000 ea (10 x bags 100 ea)
parameter
22,000 × g speed
parameter
22,000 × g speed
parameter
22,000 × g speed
parameter
22,000 × g speed
manufacturer/tradename
MTC™ Bio C2230-AM
manufacturer/tradename
MTC™ Bio C2230
manufacturer/tradename
MTC™ Bio C2231-W
manufacturer/tradename
MTC™ Bio C3150-S
sterility
sterile
sterility
sterile
sterility
sterile
sterility
sterile
material
polypropylene , amber , screw top cap, high-density polyethylene cap, self-standing
material
polypropylene , clear , blue high-density polyethylene cap, self-standing bottom, screw top cap
material
screw top cap, high-density polyethylene cap, polypropylene , clear , self-standing , blue cap
material
conical clear bottom, blue high-density polyethylene cap, screw top cap, round , polypropylene
capacity
2.0 mL
capacity
2.0 mL
capacity
2.0 mL
capacity
1.5 mL
feature
graduation
feature
graduated
feature
-
feature
-
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)