Merck
MTCR8012
MTC™ Bio IceRack™ Tube Platform with Detachable Lid
to hold, 72 x 0.5 mL tubes
Synonim(y):
centrifuge tube, cooled microtube rack, cooled tube rack, MTC Bio T8012
Informacje o tej pozycji
NACRES:
NB.12
UNSPSC Code:
41122806
eCl@ss:
32032201
Manufacturer/tradename:
MTC™ Bio R8012
Feature:
to hold 72 x 0.5 mL tubes
Material:
polypropylene
Packaging:
pack of 6 ea
Pomoc techniczna
Pozwól nam pomóc
material
polypropylene
feature
to hold 72 x 0.5 mL tubes
packaging
pack of 6 ea
manufacturer/tradename
MTC™ Bio R8012
parameter
-80-135 °C temp. range
General description
IceRack™ boxes have a removable white tube tray that allows the blue base to be filled with crushed ice or other temperature-controlling materials. They also feature a translucent cover. Temperature-sensitive samples and assays in microtubes can be processed with an extra degree of temperature stability. These boxes are suitable for use with screw-cap or snap-cap microtubes from 0.5mL to 1.5/2.0mL.
IceRack™ for 72 × 0.5mL tubes, detachable lid, tube platform, and base, numbered cavities, accepts snap cap and screw cap 0.5mL to 0.65mL tubes, 6/cs
Features and Benefits
  • Designed for crushed Ice
  • Polypropylene, -80°C to 135°C
  • Detachable “lift-off” lids
  • Economical
  • For screw cap and snap cap tubes
Legal Information
IceRack is a trademark of MTC Bio
MTC is a trademark of MTC Bio
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)