Informacje o tej pozycji
UNSPSC Code:
41103909
eCl@ss:
32110300
Manufacturer/tradename:
Eppendorf® 022638505 (NA), Eppendorf® 5804730003 (EU)
Compatibility:
for use with centrifuge models 5804/5804R/5810/5810R
Parameter:
16,400 × g max. RCF
Feature:
to hold 60 x 1.5/2.0 mL tubes, drum rotor (microtubes), non aerosol tight, without adapter, without lid
Pomoc techniczna
Pozwól nam pomócfeature
to hold 60 x 1.5/2.0 mL tubes, drum rotor (microtubes), non aerosol tight, without adapter, without lid
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 022638505 (NA), Eppendorf® 5804730003 (EU)
parameter
16,400 × g max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5804/5804R/5810/5810R
General description
A wide selections of rotors to fit the Eppendorf 5804 and 5810 centrifuges.
Legal Information
Eppendorf is a registered trademark of Eppendorf SE
Still not finding the right product?
Explore all of our products under Rotor for Eppendorf® 5804/5804R & 5810/5810R Centrifuges
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?