Nazwa produktu
Triethyl 3-methyl-4-phosphono-2-butenoate, mixture of cis and trans technical grade, 80%
Numer produktu
338958
Marka produktu
ALDRICH
Numer CAS
41891-54-7
Masa cząsteczkowa
264.26
Test | Charakterystyka techniczna |
|---|---|
| APPEARANCE | COLORLESS TO YELLOW LIQUID |
| INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
| GAS LIQUID | 78.0% (MINIMUM) AS A MIXTURE OF CIS & TRANS |
| APPROVED SEPTEMBER 22, 2008 RJM |