Nazwa produktu
Bromoacetic acid ReagentPlus®, ≥99.0%
Numer produktu
17000
Marka produktu
SIAL
Numer CAS
79-08-3
Masa cząsteczkowa
138.95
Test | Charakterystyka techniczna |
|---|---|
| APPEARANCE (COLOR) | Colorless or White to Off White |
| APPEARANCE (FORM) | Powder or Crystals or Solid or Chunks |
| TITRATION (T) NAOH 1M | 99.0 - 101.0 % |
| PURITY (GC AREA %) | ≥ 99.0 % |
| MELTING POINT | 46 - 49 C |
| SOLUBILITY (COLOR) | Colorless |
| SOLUBILITY (TURBIDITY) | Clear |
| SOLUBILITY (METHOD) | 1G IN 10ML CHLOROFORM |
| INFRARED SPECTRUM | CONFORMS TO STRUCTURE |
| PROTON NMR SPECTRUM | DIBROMOACETIC ACID ≤ 1 % |
| RECOMMENDED RETEST PERIOD | 36 MONTHS |