Nazwa produktu
2,4′-Dibromoacetophenone derivatization grade (HPLC), LiChropur™, ≥99.0% (HPLC)
Numer produktu
68082
Marka produktu
SIAL
Numer CAS
99-73-0
Masa cząsteczkowa
277.94
Test | Charakterystyka techniczna |
|---|---|
| APPEARANCE (COLOR) | White to Faint Yellow and Faint Beige and Faint Brown |
| APPEARANCE (FORM) | Powder or Crystals |
| PURITY (HPLC AREA %) | ≥ 99.0 % |
| MELTING POINT | 108 - 112 C |
| INFRARED SPECTRUM | CONFORMS TO STRUCTURE |
| PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |