Nazwa produktu
L-(+)-Arabinose BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Numer produktu
10839
Marka produktu
sigma
Numer CAS
5328-37-0
Masa cząsteczkowa
150.13
Test | Charakterystyka techniczna |
|---|---|
| APPEARANCE (COLOR) | White |
| APPEARANCE (FORM) | Powder |
| PURITY (HPLC AREA %) | ≥ 99.5 % |
| SPECIFIC ROTATION (20/D) | 102.0 - 106.0 Degrees |
| CONCENTRATION | C=10 IN WATER 24H |
| MELTING POINT | 155 - 159 C |
| LOSS ON DRYING | ≤ 0.1 % |
| RESIDUE ON IGNITION | ≤ 0.1 % |
| INFRARED SPECTRUM | CONFORMS TO STRUCTURE |
| METAL TRACE ANALYSIS (ICP) | CORRESPONDS TO REQUIREMENTS |
| ALUMINIUM (ICP) | ≤ 5 mg/kg |
| BARIUM (ICP) | ≤ 5 mg/kg |
| BISMUTH (ICP) | ≤ 5 mg/kg |
| CALCIUM (ICP) | ≤ 200 mg/kg |
| CADMIUM (ICP) | ≤ 5 mg/kg |
| COBALT (ICP) | ≤ 5 mg/kg |
| CHROMIUM (ICP) | ≤ 5 mg/kg |
| COPPER (ICP) | ≤ 5 mg/kg |
| IRON (ICP) | ≤ 5 mg/kg |
| POTASSIUM (ICP) | ≤ 50 mg/kg |
| LITHIUM (ICP) | ≤ 5 mg/kg |
| MAGNESIUM (ICP) | ≤ 5 mg/kg |
| MANGANESE (ICP) | ≤ 5 mg/kg |
| MOLYBDENUM (ICP) | ≤ 5 mg/kg |
| SODIUM (ICP) | ≤ 50 mg/kg |
| NICKEL (ICP) | ≤ 5 mg/kg |
| LEAD (ICP) | ≤ 5 mg/kg |
| STRONTIUM (ICP) | ≤ 5 mg/kg |
| ZINC (ICP) | ≤ 5 mg/kg |
| ARSENIC TRACES (MHS-AAS) | ≤ 0.1 mg/kg |
| TOTAL SULFUR AS SO4 (ICP) | ≤ 50 mg/kg |
| CHLORIDE (CL) | ≤ 50 mg/kg |
| SOLUBILITY (METHOD) | 4.50 G IN 30 ML H2O (1 M) |
| APPEARANCE (SOLUTION) | CLEAR COLORLESS |
| PH (SOLUTION) | 3.5 - 7.0 |
| RESIDUE (FILTER TEST) | NO RESIDUE |
| UV - ABS. AT 260 NM | ≤ 0.05 |
| UV - ABS. AT 280 NM | ≤ 0.04 |
| RECOMMENDED RETEST PERIOD | 72 MONTHS |