Nazwa produktu
Ammonium formate solution BioUltra, 10 M in H2O
Numer produktu
78314
Marka produktu
sigma
Numer CAS
540-69-2
Masa cząsteczkowa
63.06
Test | Charakterystyka techniczna |
|---|---|
| APPEARANCE (COLOR) | Colorless |
| APPEARANCE (FORM) | Liquid |
| TITRATION (MOLARITY) | 9.8 - 10.2 mol/l |
| DENSITY D20/4 | 1.109 - 1.149 |
| METAL TRACE ANALYSIS (ICP) | CORRESPONDS TO REQUIREMENTS |
| ALUMINIUM (ICP) | ≤ 1 mg/kg |
| BARIUM (ICP) | ≤ 1 mg/kg |
| BISMUTH (ICP) | ≤ 1 mg/kg |
| CALCIUM (ICP) | ≤ 5 mg/kg |
| CADMIUM (ICP) | ≤ 1 mg/kg |
| COBALT (ICP) | ≤ 1 mg/kg |
| CHROMIUM (ICP) | ≤ 1 mg/kg |
| COPPER (ICP) | ≤ 1 mg/kg |
| IRON (ICP) | ≤ 1 mg/kg |
| POTASSIUM (ICP) | ≤ 20 mg/kg |
| LITHIUM (ICP) | ≤ 1 mg/kg |
| MAGNESIUM (ICP) | ≤ 1 mg/kg |
| MANGANESE (ICP) | ≤ 1 mg/kg |
| MOLYBDENUM (ICP) | ≤ 1 mg/kg |
| SODIUM (ICP) | ≤ 20 mg/kg |
| NICKEL (ICP) | ≤ 1 mg/kg |
| LEAD (ICP) | ≤ 1 mg/kg |
| STRONTIUM (ICP) | ≤ 1 mg/kg |
| ZINC (ICP) | ≤ 1 mg/kg |
| ARSENIC TRACES (MHS-AAS) | ≤ 0.1 mg/kg |
| TOTAL SULFUR AS SO4 (ICP) | ≤ 50 mg/kg |
| CHLORIDE (CL) | ≤ 50 mg/kg |
| SOLUBILITY (METHOD) | NEAT |
| PH (SOLUTION) | 6.0 - 8.0 |
| RESIDUE (FILTER TEST) | NO RESIDUE |
| UV - ABS. AT 260 NM | ≤ 0.05 |
| UV - ABS. AT 280 NM | ≤ 0.04 |
| RECOMMENDED RETEST PERIOD | 48 MONTHS |