Nazwa produktu
Aldehyde Dehydrogenase from baker’s yeast ≥1 U/mg
Numer produktu
82884
Marka produktu
sigma
Numer CAS
9028-88-0
Masa cząsteczkowa
Test | Charakterystyka techniczna |
|---|---|
| APPEARANCE (COLOR) | White to Faint Brown and Faint Beige |
| APPEARANCE (FORM) | Powder or Solid or Crystals |
| SOLUBILITY (COLOR) | Colorless to Almost Colorless |
| SOLUBILITY (TURBIDITY) | Clear |
| SOLUBILITY (METHOD) | 5 MG/ML 0.1 M TRIS HCL, 0.02 % BSA, PH 8.0 |
| ENZYMATIC ACTIVITY | ≥ 1 U/mg |
| RECOMMENDED RETEST PERIOD | 48 MONTHS |