Nazwa produktu
Anti-KLF9 antibody produced in rabbit affinity isolated antibody
Numer produktu
av31369
Marka produktu
sigma
Numer CAS
Masa cząsteczkowa
Test | Charakterystyka techniczna |
|---|---|
| Concentration | 0.5-1 mg/ml |
| ELISA | Not Verified |
| Western Blotting | Antibody dilution: 1 µg/ml |
| Immunohistochemistry | Not Verified |
| Immunofluorescence | Not Verified |
| Immunoprecipitation | Not Verified |
| Flow Cytometry | Not Verified |
| Chromatin Immunoprecipitation | Not Verified |