Nazwa produktu
Anti-ZFP64 (AB1) antibody produced in rabbit IgG fraction of antiserum
Numer produktu
av39337
Marka produktu
sigma
Numer CAS
Masa cząsteczkowa
Test
Charakterystyka techniczna
Concentration0.5-1 mg/ml
ELISANot Verified
Western BlottingAntibody dilution: 1 µg/ml
ImmunohistochemistryNot Verified
ImmunofluorescenceNot Verified
ImmunoprecipitationNot Verified
Flow CytometryNot Verified
Chromatin ImmunoprecipitationNot Verified