Nazwa produktu
N-Methyl-DL-alanine
Numer produktu
m0506
Marka produktu
sigma
Numer CAS
600-21-5
Masa cząsteczkowa
103.12
Test | Charakterystyka techniczna |
|---|---|
| APPEARANCE | WHITE POWDER |
| SOLUBILITY | CLEAR COLORLESS SOLUTION AT 50 MG/ML IN WATER |
| IR SPECTRUM | CONSISTENT WITH STRUCTURE |
| WATER CONTENT BY KARL FISCHER | REPORT RESULT |
| CARBON | 44.3 - 47.8% * |
| NITROGEN | 13.2 - 13.9% * |
| SPECIFIC ROTATION | ZERO DEG (C = 1 IN WATER AT 25 DEG C) |
| PURITY BY THIN LAYER CHROMATOGRAPHY | > OR = 98% |
| * ANHYDROUS BASIS | |
| DOCUMENT # | M0506/05/12/08/1 |