Nazwa produktu
Minimum Essential Medium Eagle With Earle′s salts, non-essential amino acids and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Numer produktu
m5650
Marka produktu
sigma
Numer CAS
Masa cząsteczkowa
Test | Charakterystyka techniczna |
|---|---|
| Appearance (Turbidity) | Clear |
| Appearance (Form) | Solution |
| pH | 7.0 - 7.6 |
| Osmolality | 275 - 305 mOs/kg |
| Cell Culture Testing - MTT | Pass |
| Cell Line | Cell Line - Cell Types |
| Sterility | Pass |
| Endotoxin Level | <= 1 EU/ml |
| Glucose Concentration | 0.9 - 1.1 g/l |