Nazwa produktu
Poly(Glu, Ala, Tyr) sodium salt Glu:Ala:Tyr (6:3:1), mol wt 20,000-50,000
Numer produktu
p3899
Marka produktu
sigma
Numer CAS
97105-01-6
Masa cząsteczkowa
Test | Charakterystyka techniczna |
|---|---|
| APPEARANCE | WHITE TO OFF-WHITE POWDER OR SOLID |
| SOLUBILITY | CLEAR COLORLESS TO LIGHT YELLOW SOLUTION AT 50 MG/ML IN WATER |
| WATER CONTENT BY KARL FISCHER | < OR = 15% |
| MOLECULAR WEIGHT BY VISCOSITY | 20,000 TO 50,000 |
| DOCUMENT # | P3899/10/08/08/3 |