- Selectivity of solid phase extraction for dissolved organic matter in the hypersaline Da Qaidam Lake, China.
Selectivity of solid phase extraction for dissolved organic matter in the hypersaline Da Qaidam Lake, China.
Environmental science. Processes & impacts (2017-09-20)
Keli Yang, Yaoling Zhang, Yaping Dong, Wu Li
PMID28926062
ABSTRAKT
Dissolved organic matter (DOM) was isolated from the hypersaline Da Qaidam Lake using solid-phase extraction (SPE) methods with five different adsorbents: ENVI-Carb (non-porous graphitized carbon), HLB, PPL, and XAD-8 (polymer based), and ENVI-18 (silica based). Structure-selective assessments of SPE-DOM isolated using the different adsorbents were conducted using a combination of complementary analysis techniques, including dissolved organic carbon (DOC) and dissolved organic nitrogen (DON) analysis, as well as elemental analysis, Fourier transformed infrared spectroscopy (FTIR),
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Supel™ QuE, PSA/ENVI-Carb™ (EN) Cleanup Tube 1, pack of 100, suitable for EN 15662:2008 per BS
Supelco
Supel™ QuE, PSA/ENVI-Carb™ (EN) Cleanup Tube 2, pack of 100, suitable for EN 15662:2008 per BS
Supelco
Supel™ QuE, PSA/ENVI-Carb Tube 1, pk of 50, suitable for EN 15662:2008 per BS, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel™ QuE, PSA/ENVI-Carb Tube 2, pk of 50, suitable for EN 15662:2008 per BS, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel™ QuE, PSA/C18/ENVI-Carb Tube, pk of 50, AOAC® 2007.01, centrifuge tube volume 15 mL , Shaker Compatible