Merck
Selective TBK1/IKKi dual inhibitors with anticancer potency.
International journal of cancer (2013-10-24)
Jijia Li, Jingjia Huang, Ji-Hak Jeong, Sun-Jin Park, Rui Wei, Jieying Peng, Zhiyong Luo, Yen Ting Chen, Yangbo Feng, Jun-Li Luo
ABSTRAKT
Increasing evidence suggests that the noncanonical IKKs play critical roles in tumor genesis and development, leading to the notion that noncanonical IKKs may be good targets for cancer therapy. Here, we demonstrate that although TBK1 is not overexpressed or constitutively activated in some tumor cells, targeting IKKi induces the activation of TBK1. Therefore, simultaneously targeting both kinases is necessary to efficiently suppress tumor cell proliferation. We show that three TBK1/IKKi dual inhibitors, which are based on a structurally rigid 2-amino-4-(3'-cyano-4'-pyrrolidine)phenyl-pyrimidine scaffold, potently inhibit cell viability in human breast, prostate and oral cancer cell lines. Treatment with these TBK1/IKKi dual inhibitors significantly impairs tumor development in xenograft and allograft mouse models. The anticancer function of these inhibitors may be partially due to their suppression of TBK1/IKKi-mediated AKT phosphorylation and VEGF expression. Most importantly, these TBK1/IKKi dual inhibitors have drug-like properties including low molecular weight, low cytochrome P450 inhibition and high metabolic stability. Therefore, our studies provide proof of concept for further drug discovery efforts that may lead to novel strategies and new therapeutics for the treatment of human cancer.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Discovery® Cyano HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4 mm
Supelco
Discovery® Cyano HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Discovery® Cyano HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Discovery® Cyano Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm