Merck
Human odor detection of homologous carboxylic acids and their binary mixtures.
Chemical senses (2007-05-10)
Paul M Wise, Toshio Miyazawa, Michelle Gallagher, George Preti
ABSTRAKT
Does structural similarity of odorants influence detectability of their mixtures? To address this question, psychometric (probability of correct detection vs. concentration) functions were measured for aliphatic carboxylic acids and selected binary mixtures thereof. Unmixed stimuli included acetic (C2), butyric (C4), hexanoic (C6), and octanoic (C8) acids. Mixtures included C2 + C4, C2 + C6, and C2 + C8. Vapor-phase concentrations of individual compounds, as measured by a combination of solid-phase micro extraction and gas chromatography/mass spectrometry, were always the same, whether presented singly or in a binary mixture. Additivity of detectability was assessed with respect to response addition (independent processing of mixture components). For C2 + C6, for which the mixture components differed by 4 methylene units, and C2 + C8, which differed by 6 methylene units, response addition provided a reasonably good description of detection at all levels of performance. In contrast, for C2 + C4, which differed by only 2 methylene units, detection showed a tendency to exceed additivity at low concentrations but fell below additivity at higher concentrations. These results suggest that interaction among odors in binary mixtures does depend on structural similarity, at least for detection of carboxylic acids. Future studies can determine if this result is particular to carboxylic acids.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
SPME Fiber Assembly Kit, Assortment, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 4
Supelco
SPME Fiber Assembly Kit, Assortment #2, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #4, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #1, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #1, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #4, Fused Silica (1 cm), size 24 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 4
Supelco
SPME Fiber Assembly Kit, Assortment #1, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #2, Fused Silica (1 cm), size 24 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment, StableFlex (1 cm), needle size 24 ga, Manual Holder, pk of 4
Supelco
SPME Fiber Assembly Kit, Assortment #4, Fused Silica (1 cm), size 24 ga, Manual Holder, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #2, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #5, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 4
Supelco
SPME Fiber Assembly, 60 μm PEG, Metal Alloy (1 cm), needle size 23 ga, Manual Holder, pk of 3, purple hub
Supelco
SPME Fiber Assembly, 85 μm CAR/PDMS, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, light blue hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1cm), needle size 24 ga, Manual Holder, pk of 3, plain gray hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, blue hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 23 ga, Autosampler, pk of 3, notched gray hub
Supelco
SPME Fiber Assembly, 85 μm Polyacrylate, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, white hub
Supelco
SPME Fiber Assembly, 7 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, green hub
Supelco
SPME Fiber Assembly, 85 μm CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, light blue hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 1, pink hub
Supelco
SPME Fiber Assembly, 85 μm CAR/PDMS, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 1, light blue hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, Metal Alloy (2cm), needle size 23 ga, Autosampler, pk of 1, plain gray hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, Metal Alloy (1cm), needle size 23 ga, Autosampler, pk of, plain gray hub
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, red hub
Supelco
SPME Fiber Assembly, 85 μm Polyacrylate, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, white hub
Supelco
SPME Fiber Assembly, 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, black hub
Supelco
SPME Portable Field Sampler, coating CAR/PDMS
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, red hub