- Hydroxytyrosol inhibits hydrogen peroxide-induced apoptotic signaling via labile iron chelation.
Hydroxytyrosol inhibits hydrogen peroxide-induced apoptotic signaling via labile iron chelation.
Redox biology (2016-11-05)
Natalia Kitsati, Michalis D Mantzaris, Dimitrios Galaris
PMID27810738
ABSTRAKT
Although it is known that Mediterranean diet plays an important role in maintaining human health, the underlying molecular mechanisms remain largely unknown. The aim of this investigation was to elucidate the potential role of ortho-dihydroxy group containing natural compounds in H