- Phenformin enhances the therapeutic effect of selumetinib in KRAS-mutant non-small cell lung cancer irrespective of LKB1 status.
Phenformin enhances the therapeutic effect of selumetinib in KRAS-mutant non-small cell lung cancer irrespective of LKB1 status.
Oncotarget (2017-09-25)
Jun Zhang, Sreenivas Nannapaneni, Dongsheng Wang, Fakeng Liu, Xu Wang, Rui Jin, Xiuju Liu, Mohammad Aminur Rahman, Xianghong Peng, Guoqing Qian, Zhuo G Chen, Kwok-Kin Wong, Fadlo R Khuri, Wei Zhou, Dong M Shin
PMID28938614
ABSTRAKT
MEK inhibition is potentially valuable in targeting KRAS-mutant non-small cell lung cancer (NSCLC). Here, we analyzed whether concomitant LKB1 mutation alters sensitivity to the MEK inhibitor selumetinib, and whether the metabolism drug phenformin can enhance the therapeutic effect of selumetinib in isogenic cell lines with different LKB1 status. Isogenic pairs of KRAS-mutant NSCLC cell lines A549, H460 and H157, each with wild-type and null LKB1, as well as genetically engineered mouse-derived cell lines 634 (