Merck
  • Co-precipitation Of Phosphate And Iron Limits Mitochondrial Phosphate Availability In Saccharomyces Cerevisiae Lacking The Yeast Frataxin Homologue (YFH1).
Co-precipitation Of Phosphate And Iron Limits Mitochondrial Phosphate Availability In Saccharomyces Cerevisiae Lacking The Yeast Frataxin Homologue (YFH1).
The Journal of Thoracic and Cardiovascular Surgery (2011)
Seguin, A., et al.