Merck
8-acenaphthen-1-yl-1-phenyl-1,3,8-triaza-spiro[4.5]decan-4-one derivatives as orphanin FQ receptor agonists.
Bioorganic & medicinal chemistry letters (1999-09-07)
J Wichmann, G Adam, S Röver, A M Cesura, F M Dautzenberg, F Jenck
ABSTRAKT
A series of 8-acenaphthen-1-yl-1-phenyl-1,3,8-triaza-spiro[4.5]decan+ ++-4-one derivatives 1 was studied with respect to the binding affinity for the orphanin FQ (OFQ) and opioid (mu, kappa, delta) receptors. The influence of stereochemistry as well as the substitution pattern of the phenyl-ring in position 1 on the affinity for the orphanin FQ receptor and selectivity to opioid (mu, kappa, delta) receptors is discussed. The most interesting compound 1c was tested for its anxiolytic-like properties in vivo.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ro 65-6570 Hydrochloride, ≥98% (HPLC)