Merck
  • On-demand pulsatile intracerebral delivery of carisbamate with closed-loop direct neurostimulation therapy in an electrically induced self-sustained focal-onset epilepsy rat model.
On-demand pulsatile intracerebral delivery of carisbamate with closed-loop direct neurostimulation therapy in an electrically induced self-sustained focal-onset epilepsy rat model.
Journal of neurosurgery (2015-02-28)
Erwin Z Mangubat, Robert G Kellogg, Timothy J Harris, Marvin A Rossi
ABSTRAKT
The authors evaluated the preclinical feasibility of acutely stabilizing an active bihemispheric limbic epileptic circuit using closed-loop direct neurostimulation therapy in tandem with "on-demand'" convection-enhanced intracerebral delivery of the antiepileptic drug (AED) carisbamate. A rat model of electrically induced self-sustained focal-onset epilepsy was employed. A 16-contact depth-recording microelectrode was implanted bilaterally in the dentate gyrus (DG) of the hippocampus of Fischer 344 rats. The right microelectrode array included an integrated microcatheter for drug delivery at the distal tip. Bihemispheric spontaneous self-sustained limbic status epilepticus (SSLSE) was induced in freely moving rats using a 90-minute stimulation paradigm delivered to the right medial perforant white matter pathway. Immediately following SSLSE induction, closed-loop right PP stimulation therapy concurrent with on-demand nanoboluses of the AED [(14)C]-carisbamate (n = 4), or on-demand [(14)C]-carisbamate alone (n = 4), was introduced for a mean of 10 hours. In addition, 2 reference groups received either closed-loop stimulation therapy alone (n = 4) or stimulation therapy with saline vehicle only (n = 4). All animals were sacrificed after completing the specified therapy regimen. In situ [(14)C]-autoradiography was used to determine AED distribution. Closed-loop direct stimulation therapy delivered unilaterally in the right PP aborted ictal runs detected in either ipsi- or contralateral hippocampi. Freely moving rats receiving closed-loop direct stimulation therapy with ondemand intracerebral carisbamate delivery experienced a significant reduction in seizure frequency (p < 0.001) and minimized seizure frequency variability during the final 50% of the therapy/recording session compared with closed-loop stimulation therapy alone. Unilateral closed-loop direct stimulation therapy delivered to afferent hippocampal white matter pathways concurrent with on-demand ipsilateral intracerebral delivery of nano-bolused carisbamate can rapidly decrease the frequency of electrographic seizures in an active bihemispheric epileptic network. Additionally, direct pulsatile delivery of carisbamate can stabilize seizure frequency variability compared with direct stimulation therapy alone.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Carisbamate, ≥98% (HPLC)