Merck
  • Along with its favorable prognostic role, CLCA2 inhibits growth and metastasis of nasopharyngeal carcinoma cells via inhibition of FAK/ERK signaling.
Along with its favorable prognostic role, CLCA2 inhibits growth and metastasis of nasopharyngeal carcinoma cells via inhibition of FAK/ERK signaling.
Journal of experimental & clinical cancer research : CR (2018-02-22)
Yuan-Yuan Qiang, Chang-Zhi Li, Rui Sun, Li-Sheng Zheng, Li-Xia Peng, Jun-Ping Yang, Dong-Fang Meng, Yan-Hong Lang, Yan Mei, Ping Xie, Liang Xu, Yun Cao, Wen-Wen Wei, Li Cao, Hao Hu, Qin Yang, Dong-Hua Luo, Ying-Ying Liang, Bi-Jun Huang, Chao-Nan Qian
ABSTRAKT
CLCA2 was reported as a tumor suppressor and disregulated in breast cancer. However, its function in tumor growth and metastasis in NPC has rarely been reported. In this study, we investigated the functional and molecular mechanisms by which CLCA2 influences NPC. CLCA2 expression in human NPC cell lines and tissues was examined via real-time PCR (RT-PCR), Western blot and IHC. The biological roles of CLCA2 in proliferative, migration and invasion of NPC cell lines was evaluated in 5-8F, S18, S26 and SUNE-1 cells. Cell viability, migration and invasion were assessed in vitro by MTS, colony formation and transwell assay, respectively. CLCA2 in growth and metastasis of NPC were evaluated in vivo through NPC xenograft tumor growth, lung metastatic mice model and popliteal lymph node (LN) metastasis model. Overexpression of CLCA2 significantly decreased proliferation, migration and invasion of NPC cells. In contrast, knockdown of CLCA2 elicited the opposite effects. CLCA2 overexpression suppressed xenograft tumor growth and lung, popliteal lymph node (LN) metastasis in vivo. CLCA2 inhibited tumor metastasis through suppressing epithelial-Mesenchymal transition (EMT) and in-activating FAK/ERK1/2 signaling pathway in NPC cells. Immunohistochemical staining of 143 NPC samples revealed that CLCA2 expression was an independent, favorable prognostic factor for overall survival and distant metastasis-free survival of patients. In addition, inhibition of FAK and ERK1/2 reversed CLCA2 silencing-induced tumor cell migration. Furthermore, inhibitors against chloride channels suppressed NPC cellular migration which could have been enhanced by the presence of CLCA2. CLCA2 suppress NPC proliferation, migration, invasion and epithelial-mesenchymal transition through inhibiting FAK/ERK signaling.