Merck
Characterization of PIK3CA and PIK3R1 somatic mutations in Chinese breast cancer patients.
Nature communications (2018-04-11)
Li Chen, Liu Yang, Ling Yao, Xia-Ying Kuang, Wen-Jia Zuo, Shan Li, Feng Qiao, Yi-Rong Liu, Zhi-Gang Cao, Shu-Ling Zhou, Xiao-Yan Zhou, Wen-Tao Yang, Jin-Xiu Shi, Wei Huang, Xin Hu, Zhi-Ming Shao
ABSTRAKT
Deregulation of the phosphoinositide 3-kinase (PI3K) pathway contributes to the development and progression of tumors. Here, we determine that somatic mutations in PIK3CA (44%), PIK3R1 (17%), AKT3 (15%), and PTEN (12%) are prevalent and diverse in Chinese breast cancer patients, with 60 novel mutations identified. A high proportion of tumors harbors multiple mutations, especially PIK3CA plus PIK3R1 mutations (9.0%). Next, we develop a recombination-based mutation barcoding (ReMB) library for impactful mutations conferring clonal advantage in proliferation and drug responses. The highest-ranking PIK3CA and PIK3R1 mutations include previously reported deleterious mutations, as well as mutations with unknown significance. These PIK3CA and PIK3R1 impactful mutations exhibit a mutually exclusive pattern, leading to oncogenesis and hyperactivity of PI3K pathway. The PIK3CA impactful mutations are tightly associated with hormone receptor positivity. Collectively, these findings advance our understanding of PI3K impactful mutations in breast cancer and have important implications for PI3K-targeted therapy in precision oncology.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-HA antibody, Mouse monoclonal, clone HA-7, purified from hybridoma cell culture